missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ M871

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
M871 is a selective galanin GAL2 receptor antagonist (Ki values are 13.1 and 420 nM for GAL2 and GAL1 receptors respectively). Blocks the pro-nociceptive effect of GAL2 receptor agonists.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction