missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ α-MSH
GREENER_CHOICE

Product Code. 18767227
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18767227

Brand: Tocris Bioscience™ 2584/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous melanocortin receptor agonist

α-MSH is an endogenous melanocortin receptor agonist (Ki values are 0.12, 31, 660 and 5700 nM for MC1, MC3, MC4 and MC5 receptors respectively). Anti-inflammatory peptide; antagonizes proinflammatory mediators, including TNF-α, IL-6 and NO and induces anti-inflammatory cytokine IL-10. Inhibits food intake and induces penile erections following i.c.v. administration.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.