Learn More
Tocris Bioscience™ Pepstatin A
GREENER_CHOICE
Product Code. 18064772
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 18064772
Brand: Tocris Bioscience™ 1190/10
Selective and high affinity cathepsin D inhibitor
Pepstatin A is an irreversible inhibitor of aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d (Cat. No. 4545).
TRUSTED_SUSTAINABILITY
Specifications
Molecular Weight (g/mol) Observed M.W.: 685.91; Theoretical M.W.: 0
Quantity 10 mg