Learn More
Tocris Bioscience™ Pepstatin A
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Pepstatin A is an irreversible inhibitor of aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d (Cat. No. 4545).
Specifications
Specifications
| Molecular Weight (g/mol) | Observed M.W.: 685.91; Theoretical M.W.: 0 |
| Quantity | 10 mg |