Learn More
Tocris Bioscience™ ProTx II
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
missing translation for 'unitSize'
100µg
Description
ProTx II is a selective NaV1.7 channel blocker. Shifts activation gating positively and decreases current magnitude. Displays 100-fold selectivity over other sodium channel subtypes.
Specifications
Specifications
| Quantity | 100 μg |