missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Secretin (human)

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Secretin (human) is a gastrointestinal peptide that stimulates pancreatic and biliary secretion. Also thought to play a role in the regulation of the hypothalamus-pituitary-adrenal axis.
Secretin (rat) also available.
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction