Learn More
Tocris Bioscience™ Senktide
GREENER_CHOICE
Product Code. 18757716
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. 18757716
Brand: Tocris Bioscience™ 1068/500U
Tachykinin NK3 agonist
Senktide is a NK3 tachykinin receptor agonist. Causes direct excitation of dopamine neurons; enhances dopaminergic function. Induces locomotor activity.
TRUSTED_SUSTAINABILITY
Specifications
Purity 95%
Content And Storage Store at -20°C
Quantity 500 μg