Learn More
TOE1 Antibody (1C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00114034-M02
Product Code. 18473889
  • € 589.00 / 0.10mg
Description
TOE1 Monoclonal antibody specifically detects TOE1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TOE1
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
FLJ13949, target of EGR1 protein 1, target of EGR1, member 1 (nuclear)
TOE1 (AAH09364, 1 a.a. ∽ 510 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAADSDDGAVSAPAASDGGVSKSTTSGEELVVQVPVVDVQSNNFKEMWPSLLLAIKTANFVAVDTELSGLGDRKSLLNQCIEERYKAVCHAARTRSILSLGLACFKRQPDKGEHSYLAQVFNLTLLCMEEYVIEPKSVQFLIQHGFNFNQQYAQGIPYHKGNDKGDESQSQSVRTLFLELIRARRPLVLHNGLIDLVFLYQNFYAHLPESLGTFTADLCEMFPAGIYDTKYAAEFHARFVASYLEYAFRKCERENGKQRAAGSPHLTLEFCNYPSSMRDHIDYRCCLPPATHRPHPTSICDNFSAYGWCPLGPQCPQSHDIDLIIDTDEAAAEDKRRRRRRREKRKRALLNLPGTQTSGEAKDGPPKKQVCGDSIKPEETEQEVAADETRNLPHSKQGNKNDLEMGIKAARPEIADRATSEVPGSQASPNPVPGDGLHRAGFDAFMTGYVMAYVEVSQGPQPCSSGPWLPECHNKVYLSGKAVPLTVAKSQFSRSSKAHNQKMKLTWGSS
0.1 mg
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
1C12
Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
AAH09364
Mouse
IgG purified
RUO
114034
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG1 κ