missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOE1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04022AF750
This item is not returnable.
View return policy
Description
TOE1 Polyclonal antibody specifically detects TOE1 in Human samples. It is validated for Western Blot
Specifications
| TOE1 | |
| Polyclonal | |
| Western Blot | |
| FLJ13949, target of EGR1 protein 1, target of EGR1, member 1 (nuclear) | |
| The immunogen recognized by this antibody maps to a region between residue 460 and 510 of human Target of EGR1, Member 1 using the numbering given in entry NP_079353.3 (GeneID 114034). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 114034 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction