missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tollip Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77255UV
This item is not returnable.
View return policy
Description
Tollip Polyclonal antibody specifically detects Tollip in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Tollip | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human TOLLIP. The immunogen is located within amino acids 70 - 120 of TOLLIP. Amino Acid Squence: ETPTAHNGAKNPRWNK | |
| 0.1 mL | |
| Signal Transduction | |
| 54472 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction