missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tollip Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76681DL594
This item is not returnable.
View return policy
Description
Tollip Polyclonal antibody specifically detects Tollip in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Tollip | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human TOLLIP. The immunogen is located within the last 50 amino acids of TOLLIP. Amino Acid Squence: RSVLEAQRGNKDAAINS | |
| 0.1 mL | |
| Signal Transduction | |
| 54472 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction