missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™

Catalog Number p-7073621
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
Melting Point -95°C
Color Colorless
Boiling Point 111°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Formula Weight 92.14g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'