missing translation for 'onlineSavingsMsg'
Learn More

Toluene-d8, 99.6%(Isotopic), Thermo Scientific Chemicals

Product Code. 11327938
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327938 1 g 1g
11337938 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11327938 Supplier Thermo Scientific Alfa Aesar Supplier No. 036477.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Toluene is mainly used as a precursor to benzene. Paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, and leather tanners. And also used as an octane booster in gasoline fuels used in internal combustion engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
MDL Number MFCD00044638
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
Synonym toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
PubChem CID 74861
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
SMILES CC1=CC=CC=C1

Specifications

Physical Form Liquid
Density 0.94g/mL
Flash Point 10° (50°F)
Odor Aromatic
Quantity 1 g
Melting Point -93°C
Boiling Point 111°C
UN Number UN1294
Formula Weight 100.19
Percent Purity 99.6%
Chemical Name or Material Toluene-d8, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.