Learn More
Tom22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11278-1-AP-20UL
 View more versions of this product
Product Code. 16868243
  • € 145.00 / 20µL
Description
Tom22 has a negatively charged N-terminal region exposed to the cytosol, a putative transmembrane region, and a C-terminal intermembrane space region with little negative charge. The following functions can be assigned to the three domains of Tom22: the cytosolic N-terminal domain plays a dual role, specifically important for presequence binding; the intermembrane space domain provides a trans binding site for presequences, and the single membrane anchor of Tom22 is crucial for the integrity of the GIP (general import gene) complex. The TOM complex of mammalian mitochondria resembles the fungal Tom complex, but is distinct from the plant TOM system. Thus, while unique components of the mammalian mitochondrial import system have been identified (e. g. TOM34 and metaxin), Tom22, and Tom37 have not been identified in plant mitochondria.
Specifications
Specifications
Tom22
Polyclonal
Unconjugated
TOMM22
1C9 2, hTom22, MST065, MSTP065, TOM22, TOMM22
Rabbit
Antigen Affinity Chromatography
RUO
223696, 300075, 56993
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q75Q41, Q9CPQ3, Q9NS69
TOMM22
Tom22 Fusion Protein Ag1797
20 μL
Primary
Mouse, Rat, Human