missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOM70 Polyclonal antibody specifically detects TOM70 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | TOM70 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast) |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 245-608 of human TOM70 (NP_055635.3). KNREPLMPSPQFIKSYFSSFTDDIISQPMLKGEKSDEDKDKEGEALEVKENSGYLKAKQYMEEENYDKIISECSKEIDAEGKYMAEALLLRATFYLLIGNANAAKPDLDKVISLKEANVKLRANALIKRGSMYMQQQQPLLSTQDFNMAADIDPQNADVYHHRGQLKILLDQVEEAVADFDECIRLRPESALAQAQKCFALYRQAYTGNNSSQIQAAMKGFEEVIKKFPRCAEGYALYAQALTDQQQFGKADEMYDKCIDLEPDNATTYVHKGLLQLQWKQDLDRGLELISKAIEIDNKCDFAYETMGTIEVQRGNMEKAIDMFNKAINLAKSEMEMAHLYSLCDAAHAQTEVAKKYGLKPPTL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?