missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM20 Antibody (1B2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60248CL1
This item is not returnable.
View return policy
Description
TOMM20 Monoclonal antibody specifically detects TOMM20 in Human,Mouse samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TOMM20 | |
| Monoclonal | |
| CoraFluor 1 | |
| KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II | |
| Recombinant human Tomm20 (25-145aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 9804 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1B2 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction