missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM34 Antibody (Tomm34-4.1), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11704C
This item is not returnable.
View return policy
Description
TOMM34 Monoclonal antibody specifically detects TOMM34 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| TOMM34 | |
| Monoclonal | |
| DyLight 650 | |
| hTom34, HTOM34P, mitochondrial import receptor subunit TOM34, outer mitochondrial membrane translocase (34kD), TOM34, Translocase of outer membrane 34 kDa subunit, translocase of outer mitochondrial membrane 34, URCC3 | |
| Recombinant human Tomm34 | |
| 0.1 mL | |
| Signal Transduction | |
| 10953 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Tomm34-4.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction