missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM34 Monoclonal Antibody (OTI2D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503184S
This item is not returnable.
View return policy
Description
TOMM34 Monoclonal antibody specifically detects TOMM34 in Human samples. It is validated for Western BlotSpecifications
| TOMM34 | |
| Monoclonal | |
| 0.79 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15785 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10953 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2D8 | |
| Unconjugated | |
| Tomm34 | |
| 2610100K07Rik; hTom34; HTOM34P; mitochondrial import receptor subunit TOM34; outer mitochondrial membrane translocase (34kD); TOM34; Tomm34; Translocase of outer membrane 34 kDa subunit; translocase of outer mitochondrial membrane 34; URCC3 | |
| Full length human recombinant protein of human TOMM34 produced in HEK293 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction