missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP1 Monoclonal Antibody (23B11)

Product Code. 10617933
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10617933 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10617933 Supplier Invitrogen™ Supplier No. 44321M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TOP1 Monoclonal Antibody for Western Blot

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP1
Applications Western Blot
Classification Monoclonal
Clone 23B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with PEG, sucrose and 0.09% sodium azide; pH 7.3
Gene Top1
Gene Accession No. P11387
Gene Alias Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase
Gene Symbols Top1
Host Species Mouse
Immunogen Peptide containing amino acid residues 699-725, conjugated to KLH.
Purification Method Size-exclusion chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7150
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.