missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1 Monoclonal Antibody (OTI3B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809934
This item is not returnable.
View return policy
Description
Topoisomerase I encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.Specifications
| TOP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11387 | |
| TOP1 | |
| Human recombit protein fragment corresponding to amino acids 93-151 of human TOP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B9 | |
| Unconjugated | |
| TOP1 | |
| TOPI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7150 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction