missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578203
This item is not returnable.
View return policy
Description
Positive Control: 293T, 293T NE Predicted Reactivity: Rat (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
Specifications
| TOP1 | |
| Polyclonal | |
| Unconjugated | |
| Top1 | |
| Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64550, 7150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P11387, Q9WUL0 | |
| Top1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Topoisomerase I. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction