missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP1 Recombinant Human Monoclonal Antibody (K94015_6C5), Invitrogen™

Product Code. 30210608
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30210608 50 μL 50µg
30207537 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30210608 Supplier Invitrogen™ Supplier No. MA555613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Product is compatible with indirect ELISA (1 μg/mL, capture antibody). Sequence of this protein is as follows: EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone K94015_6C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Top1
Gene Accession No. P11387
Gene Alias Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase
Gene Symbols Top1
Host Species Human
Immunogen Recombinant human TOP1 (immunogen range is 221-420).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.