missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1MT Antibody (TOP1MT/568), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08305AF532
This item is not returnable.
View return policy
Description
TOP1MT Monoclonal antibody specifically detects TOP1MT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TOP1MT | |
| Monoclonal | |
| Alexa Fluor 532 | |
| mitochondrial, mitochondrial DNA topoisomerase I, topoisomerase (DNA) I, mitochondrial | |
| Recombinant full-length human TOP1MT protein (Uniprot: Q969P6) | |
| 100 μg | |
| Cancer | |
| 116447 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| TOP1MT/568 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction