missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2A Antibody (OTI2A9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74558H
This item is not returnable.
View return policy
Description
TOP2A Monoclonal specifically detects TOP2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TOP2A | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TOP2A | |
| Recombinant protein fragment corresponding to amino acids 1100-1531 of human TOP2A (NP_001058) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2A9 | |
| Western Blot, Immunohistochemistry | |
| DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Phospho Specific, Stem Cell Markers | |
| 7153.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction