missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2A Antibody (TOP2A/1361), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54546F
This item is not returnable.
View return policy
Description
TOP2A Monoclonal antibody specifically detects TOP2A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| TOP2A | |
| Monoclonal | |
| FITC | |
| DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A | |
| Recombinant human TOP2A fragment (aa1352-1493) (Uniprot: P11388) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 7153 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| TOP2A/1361 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction