missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP2A Monoclonal Antibody (A3F12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544952
This item is not returnable.
View return policy
Description
TOP2A Monoclonal Antibody for Western Blot, IHC (P)
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2 and M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
Specifications
| TOP2A | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P11388, Q01320 | |
| TOP2A | |
| Synthetic peptide within human Topoisomerase II alpha aa1480-1531. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| A3F12 | |
| Unconjugated | |
| TOP2A | |
| DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase 2-alpha-like protein; DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2; Top-2; Top2a; TOPOII; topoisomerase (DNA) 2 alpha; topoisomerase (DNA) II alpha; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha; topoisomersae II; TP2A | |
| Mouse | |
| Protein G | |
| RUO | |
| 21973, 7153 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction