missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116710
This item is not returnable.
View return policy
Description
Positive Samples: Mouse testis Immunogen sequence: MILARNYLDV YPYDHWSDKI LPVYEQGSHF QPSTVEMVDG ETSPPKLLTE ADLIALMEKH GIGTDATHAE HIETIKARMY VGLTPDKRFL PGHLGMGLVE.
DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I (Topo I) and Topo II relax both positive and negative supercoils, Topo III relaxes only negative supercoils. Topo IIIalpha exists as a long and a short isoform, which are produced by alternative splicing. Topo IIIalpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. Topo IIIbeta exists as three isoforms,beta-1,beta -2 and beta-3, also produced by alternative splicing. Topo IIIbeta-1 is expressed in testis, heart, and skeletal muscle, whereas Topo IIIbeta-2 is expressed in thymus, kidney and pancreas.
Specifications
| TOP3A | |
| Polyclonal | |
| Unconjugated | |
| Top3a | |
| DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; TOP3; TOP3A; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; ZGRF7; zinc finger, GRF-type containing 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21975 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.46 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O70157 | |
| Top3a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TOP3A (XP_0115223031). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction