missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Topiramate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Topiramate is an anticonvulsant. Antagonizes GluK1 (formerly GluR5) kainate receptors (IC50 = 0.46 μM), acts as a positive allosteric modulator of GABAA receptor-mediated currents, inhibits Nav channels (IC50 = 48.9 μM) and inhibits L-type Ca2+ channels. Also inhibits carbonic anhydrase (CA) (Ki values are 0.1 and 0.2 μM at rat CA II and CA IV respectively), which lowers intracellular neuronal pH.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Topiramate |
| Purity | >99% |
| Molecular Weight (g/mol) | 339.36 |
| Inhibitors | Topiramate |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction