missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR1AIP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98140-50ul
This item is not returnable.
View return policy
Description
TOR1AIP2 Polyclonal antibody specifically detects TOR1AIP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TOR1AIP2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| FLJ77012, IFRG15, interferon responsive gene 15, LULL1MGC120077, lumenal domain like LAP1, Lumenal domain-like LAP1, MGC120074, MGC120075, MGC120076, MGC126581, MGC138430, NET9, RP11-12M5.5, torsin A interacting protein 2, torsin-1A-interacting protein 2 | |
| Produced in rabbits immunized with E. coli-derived Human TOR1AIP2 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 163590 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction