Learn More
Invitrogen™ TOR3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540655
This item is not returnable.
View return policy
Description
Peptide sequence: FHFPHPKYVD LYKEQLMSQI RETQQLCHQT LFIFDEAEKL HPGLLEVLGP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 83%.
The function remains unknown.
Specifications
| TOR3A | |
| Polyclonal | |
| Unconjugated | |
| TOR3A | |
| Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64222 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H497 | |
| TOR3A | |
| Synthetic peptide directed towards the middle region of human TOR3A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |