missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC3/CRTC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46832
This item is not returnable.
View return policy
Description
TORC3/CRTC3 Polyclonal specifically detects TORC3/CRTC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TORC3/CRTC3 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| CRTC3 | |
| The immunogen for this product maps to a region between residue 569 and 619 of human transducer of regulated CREB protein 3 using the numbering given in entry NP_073606.3 (GeneID 64784). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 64784 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction