missing translation for 'onlineSavingsMsg'
Learn More

SCA Hygiene Products™ Tork Plastic Centrefeed M2 Dispenser

Product Code. 10625685 Shop All Tork Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10625685 Black Each
10615685 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625685 Supplier SCA Hygiene Products™ Supplier No. 559008_CLEAN

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Centrefeed Roll System M2 Dispenser
Color Black
Depth (Metric) 227 mm
Width (Metric) 239 mm
Height (Metric) 360 mm
Material Plastic
For Use With (Equipment) Tork Centrefeed Wiping Paper Roll
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.