Learn More
DiversifiedBiotech Tough-Spots™
Description
- Circular cryogenic labels formatted for handwriting
- Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
- Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job!
- Adheres to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling or degrading
- Flexible polyolefin material and acrylic adhesive expand and contract with temperature fluctuations
- Roll of 1007
Specifications
Specifications
| Adhesive Type | Peel and Stick, Acrylic |
| Color | White |
| Finish | Semi-gloss |
| Format | Roll |
| Material | Polyolefin |
| Print Technology | Laser |
| Product Type | Ultra Reflective Letter Label |
| Sterility | Non-sterile |
| Quantity | 1500/Pk. |
| Length (English) | 0.1875 in. |
| Show More |