Learn More
Brady™ ToughWash™ Metal Detectable, Washdown Resistant Polyester Label Tape for M7 Printers
ToughWash™ polyester film with acrylic adhesive is a strong, print-on-demand label specially formulated for the Food and Beverage Processing industry.
Brand: Brady™ 173469
This item is not returnable.
View return policy
Description
- Capable of being detected by commercially available metal detectors
- Label construction is tough against harsh washdown environments and withstands washdowns up to 175°F (80°C) and pressure up to 1500 PSI
- Service temperature of -40°C to +110°C (-40°F to +230°F)
- Made of B-854 polyester material
Specifications
| Not Autoclavable | |
| None | |
| Matte | |
| Polyester | |
| Thermal Transfer | |
| Non-sterile | |
| M710, BMP71 | |
| 75 ft. | |
| -40°F to +230°F | |
| 0.009 in. | |
| 75 ft. x 2 in. | |
| 22.86 m | |
| 50.8 mm |
| Metallic | |
| Yellow | |
| Continuous | |
| Single-side Printable | |
| Label | |
| Facility Identification, Food and Beverage Process Labeling, General Identification, Safety Identification | |
| No | |
| Rectangle | |
| -40°C to +110°C | |
| 0.229 mm | |
| 75 ft. | |
| 2 in. | |
| BM7C-2000-854-YL |