missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOX2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94397-0.1ml
This item is not returnable.
View return policy
Description
TOX2 Polyclonal antibody specifically detects TOX2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TOX2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C20orf100, chromosome 20 open reading frame 100, dJ495O3.1, GCX1, GCX-1dJ1108D11.2, granulosa cell HMG box 1, Granulosa cell HMG box protein 1, granulosa cell HMG-box protein 1, MGC15880, TOX high mobility group box family member 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human TOX2 (NP_116272.1). MSDGNPELLSTSQTYNGQSENNEDYEIPPITPPNLPEPSLLHLGDHEASYHSLCHGLTPNGLLPAYSYQAMDLPAIMVSNMLAQDSHLLSGQLPTIQEMVHSEVAAYDSGRPGPLLGRPAMLASHMSALSQSQLISQMGI | |
| 0.1 mL | |
| Endocrinology, Neuroscience | |
| 84969 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction