Learn More
TOX2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30474-1-AP-20UL
This item is not returnable.
View return policy
Description
The TOX2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TOX2. This antibody recognizes human, mouse, rat antigen. The TOX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TOX2 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| C20orf100, dJ1108D11.2, dJ495O3.1, GCX 1, GCX1, TOX2 | |
| 20 μL | |
| 84969 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |