Learn More
TOX2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30474-1-AP-20UL
 View more versions of this product
Product Code. 18034825
  • € 145.00 / 20µL
Description
The TOX2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TOX2. This antibody recognizes human, mouse, rat antigen. The TOX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TOX2
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
C20orf100, dJ1108D11.2, dJ495O3.1, GCX 1, GCX1, TOX2
20 μL
84969
Store at -20°C. Stable for one year after shipment.