missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxoplasma Gondii P30 Antibody (P30/3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66486JF646
This item is not returnable.
View return policy
Description
Toxoplasma Gondii P30 Monoclonal specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| ToxoplasmaGondiiP30 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Protozoa, T. gondii | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| P30/3 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| p30, SAG1, SAG1 protein | |
| Purified T. gondii P30 antigen | |
| 0.1 mL | |
| Specific for the Toxoplasma gondii P30 membrane protein. | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction