missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TP 238

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
TP 238 is a cat eye syndrome chromosome region, candidate 2 (CECR2) and bromodomain PHD finger transcription factor (BPTF/FALZ) inhibitor (IC50 values are 10-30 and 100-350 nM, respectively).
A concentration of no more than 2 μM is recommended for use in cellular assays.
Specifications
Specifications
| Chemical Name or Material | N-[(3-(1 H)-Pyrazol-1-yl)propyl)-6-(4-(3-(dimethylamino)propoxy)phenyl)-2-(methylsulfonyl)pyrimidin-4-amine hydrochloride |
| CAS | 2415263-05-5 |
| Quantity | 10 mg |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C22H30N6O<sub>3S.HCl |
| Formula Weight | 2576 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction