missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53BP1 Monoclonal Antibody (OTI9C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807214S
This item is not returnable.
View return policy
Description
TP53BP1 Monoclonal antibody specifically detects TP53BP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TP53BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15116 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9C8 | |
| Unconjugated | |
| Pdcd1 | |
| 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1 | |
| Full length human recombinant protein of human PDCD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction