missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53BP1 Monoclonal Antibody (OTI9C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807214
This item is not returnable.
View return policy
Description
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.Specifications
| TP53BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12888 | |
| TP53BP1 | |
| Human recombit protein fragment corresponding to amino acids 1-300 of human TP53BP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9C8 | |
| Unconjugated | |
| TP53BP1 | |
| 53BP1, TDRD30, TP53, p202, p53BP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction