Learn More
TP53BP1 Mouse anti-Human, Clone: OTI6D9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806950
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.Specifications
| TP53BP1 | |
| Monoclonal | |
| Unconjugated | |
| TP53BP1 | |
| 53BP1, TDRD30, TP53, p202, p53BP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6D9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q12888 | |
| TP53BP1 | |
| Human recombit protein fragment corresponding to amino acids 1-300 of human TP53BP1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.