Learn More
Invitrogen™ TP53I11 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599198
Product Code. 16683445
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total TP53I11.
TP53I11 is a 177 amino acid tumor suppressor belonging to the p53-induced protein gene (PIG) family. The PIG gene family encodes redox-controlling proteins that are involved in p53 tumor suppressor activity. It is suggested that PIG11 is involved in arsenic trioxide As(2)O(3)-induced apoptosis in certain cell lines and may play a significant role in tumor suppression through promotion of cell apoptosis. The gene encoding PIG11 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TP53I11
Polyclonal
Unconjugated
TP53I11
p53-induced gene 11 protein; Pig11; TP53I11; transformation related protein 53 inducible protein 11; Trp53i11; tumor protein p53 inducible protein 11; tumor protein p53-inducible protein 11
Rabbit
Affinity chromatography
RUO
311209, 9537
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
B3DMA0, O14683
TP53I11
A synthesized peptide derived from human TP53I11(Accession O14683), corresponding to amino acid residues M10-L60.
100 μL
Primary
Human, Rat