missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53RK, Mouse anti-Human, Clone: 2E8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00112858-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: HDEDLIHGDLTTSNMLLKPPLEQLNIVLIDFGLSFISALPEDKGVDLYVLEKAFLSTHPNTETVFEAFLKSYSTSSKKARPVLKKLDEVRLRGRKRSMVGSpecifications
| TP53RK | |
| Monoclonal | |
| Unconjugated | |
| TP53 regulating kinase | |
| BUD32/C20orf64/Nori-2/Nori-2p/PRPK/dJ101A2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2E8 | |
| In 1x PBS, pH 7.4 | |
| BC009727 | |
| TP53RK | |
| TP53RK (AAH09727, 154 a.a. ∼ 253 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 112858 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction