missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TP53TG5 Recombinant Protein Antigen

Product Code. 18651354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18651354 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18651354 Supplier Novus Biologicals™ Supplier No. NBP321410PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 27296
Molecular Weight (g/mol) MolecularWeight-theroretical: 26 kDa
Quantity 100 μL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias C20orf10, chromosome 20 open reading frame 10, CLG01, dJ453C12.5, TP53 target 5, TP53-inducible gene 5 protein, TP53-target gene 5 protein
Gene Symbol TP53TG5
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-21410. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Research Category Signal Transduction
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein TP53TG5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.