missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP63 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30824.100ug
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against partial recombinant human TP63.
- High specificity
- High reactivity
Specifications
| TP63 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant human TP63. | |
| In 10 mM PBS. | |
| AIS/B(p51A)/B(p51B)/EEC3/KET/LMS/NBP/OFC8/RHS/SHFM4/TP53CP/TP53L/TP73L/p40/p51/p53CP/p63/p73H/p73L | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 8626 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Q9H3D4 | |
| TP63 | |
| Recombinant protein corresponding to human TP63. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction