missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP73-AS1 Antibody (OTI2B9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74559R
This item is not returnable.
View return policy
Description
TP73-AS1 Monoclonal specifically detects TP73-AS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TP73-AS1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TP73-AS1 | |
| Full length recombinant protein of human TP73-AS1 (NP_997198) | |
| 0.1 mL | |
| 57212.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| KIAA0495, MGC138189, PDAM | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction