missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52 Rabbit anti-Human, Polyclonal , Abnova™
TPD52 polyclonal antibody raised against recombinant human TPD52.
Brand: Abnova PAB29480.100uL
This item is not returnable.
View return policy
Description
Sequence: KSFEEKVENLKSKVGGTKPAGGDFGEVLNSAANASATTTEPLPEKTQESLSpecifications
| TPD52 | |
| Polyclonal | |
| TPD52 polyclonal antibody raised against recombinant human TPD52. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| TPD52 | |
| Recombinant protein corresponding to amino acids of human TPD52. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| D52/N8L/PC-1/PrLZ/hD52 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7163 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction