missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPD52L2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518797
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications
| TPD52L2 | |
| Polyclonal | |
| Unconjugated | |
| Tpd52l2 | |
| 2810411G23Rik; AU016537; C86069; D54; DKFZp686A1765; HCCR-binding protein 2; hD54; Tpd52l2; TPD54; tumor protein D52 like 2; tumor protein D52-like 2; Tumor protein D54 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 7165 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43399 | |
| Tpd52l2 | |
| Synthetic peptide sequence (SGDKPLSDPAP) corresponding to the C-terminus amino acids of TPD52L2 (aa 195-206). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction