Learn More
TPD52L3 Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504953
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Belongs to the TPD52 family and is specifically expressed in testis. Expressed at 5.6-fold higher levels in adult testis than in fetal testis.Specifications
| TPD52L3 | |
| Monoclonal | |
| Unconjugated | |
| TPD52L3 | |
| NYDSP25, hD55 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89882 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2D2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96J77 | |
| TPD52L3 | |
| Full length human recombit protein of human TPD52L3 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.