missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPMT Antibody (2E7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97939H
This item is not returnable.
View return policy
Description
TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, ELISA.
Specifications
| TPMT | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TPMT | |
| Recombinant human TPMT (1-245aa) purified from E. coli | |
| 100 μL | |
| Proteases & Other Enzymes, Signal Transduction | |
| 7172 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 20000000 | |
| Western Blot, ELISA | |
| EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human TPMT mAb, clone 2E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TPMT protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction