missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPMT Antibody (OTI2A2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74570CL1
This item is not returnable.
View return policy
Description
TPMT Monoclonal antibody specifically detects TPMT in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TPMT | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase | |
| Full length human recombinant protein of human TPMT (NP_000358) produced in HEK293T cell. | |
| 0.1 mL | |
| Proteases & Other Enzymes, Signal Transduction | |
| 7172 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2A2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction